Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ThioUrea, ≥99.999% (metals basis), MilliporeSigma™ Supelco™

Catalog No. 111018362 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8362 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8362 Supplier MilliporeSigma Supelco Supplier No. 308855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00008067
Synonym Sulfourea; Thiocarbamide

Specifications

Melting Point 170°C to 176°C
CAS 62-56-6
Linear Formula NH2 CSNH2
Quantity 5 g
Percent Purity ≥99.999% (metals basis)
RTECSNumber YU2800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.