Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiourea, 99+%, for analysis

Catalog No. AC220050010
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220050010 1 kg Plastic bottle
AC220052500 250 g Plastic bottle
AC220050050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC220050010 Supplier Thermo Scientific Chemicals Supplier No. 220050010
Add to Cart

CAS: 62-56-6 | CH4N2S | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 171.0°C to 175.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Iodimetry)
Packaging Plastic bottle
Linear Formula H2NCSNH2
Quantity 1 kg
Beilstein 03, 180
Fieser 01,1164; 02,412; 03,290; 06,586; 11,519; 15,308
Merck Index 15, 9521
Solubility Information Solubility in water: 136g/L (20°C). Other solubilities: 12 g/100g methanol (20°C), soluble in isopropylalcohol, butanol , pentanol, sparingly soluble in ether
Formula Weight 76.12
Percent Purity 99+%
Grade Analysis
Physical Form Crystals
Chemical Name or Material Thiourea, For analysis
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep
EINECSNumber 200-543-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.