Learn More
METTLER TOLEDO 3P INDUSTRIAL THIOUREA CLEANING SOLN 250ML
Catalog No. NC0829290
Encompass_Preferred
Thiourea solution for cleaning junctions with silver sulfide contamination. Treatment until discoloration.
Catalog No. NC0829290 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51350102
Thiourea solution for cleaning junctions with silver sulfide contamination. Treatment until discoloration.