Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiourea, n-[(1,2-dihydro-6,8-dimethyl-2-oxo-3-quinolinyl)methyl]-n'-(4-ethoxyphenyl) | 484006-66-8 | 98.2% | 425.54 g/mol | C23H27N3O3S | 1 ML
SDP

Catalog No. 5000247216
Encompass_Preferred

β-Glucuronidase-IN-1 is a potent, selective, uncompetitive inhibitor of E. coli β-glucuronidase with reported IC50 of 283 nM and Ki of 164 nM. The compound is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid material in multiple sizes, and is intended for biochemical and in vivo research applications.

  • Potent, selective, uncompetitive inhibitor of bacterial β-glucuronidase
  • Reported IC50 283 nM and Ki 164 nM
  • Supplied as 10 mM solution in DMSO (1 mL) or as solid in multiple pack sizes
  • Molecular formula C23H27N3O3S; molecular weight 425.54 g/mol
  • High purity (≈98.2%) suitable for biochemical studies
  • Certificate of analysis, datasheet, and SDS available

Catalog No. 50-002-47216 Supplier Medchemexpress LLC Supplier No. HY10308110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Glucuronidase-IN-1 is a potent, selective, uncompetitive inhibitor of E. coli β-glucuronidase with reported IC50 of 283 nM and Ki of 164 nM. The compound is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid material in multiple sizes, and is intended for biochemical and in vivo research applications.

  • Potent, selective, uncompetitive inhibitor of bacterial β-glucuronidase
  • Reported IC50 283 nM and Ki 164 nM
  • Supplied as 10 mM solution in DMSO (1 mL) or as solid in multiple pack sizes
  • Molecular formula C23H27N3O3S; molecular weight 425.54 g/mol
  • High purity (≈98.2%) suitable for biochemical studies
  • Certificate of analysis, datasheet, and SDS available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.