Learn More
Medchemexpress LLC Thiourea, N-phenyl- | 103-85-5 | MFCD00004933 | 99.3% | 152.22 g/mol | C7H8N2S | 50g
SDP
Catalog No. 5000268274
Encompass_Preferred
Phenylthiourea (Phenylthiocarbamide) is an inhibitor for phenoloxidase Phenylthiourea inhibits enzymatic oxidation of DOPA by phenoloxidase (Ki 0 21 M) Phenylthiourea is an effective inhibitor for tyrosinase Phenylthiourea can lead to graying of hair in black rats due to the interference with melanin formation[1][2]
Catalog No. 50-002-68274 Supplier Medchemexpress LLC Supplier No. HYY035150G
Phenylthiourea (Phenylthiocarbamide) is an inhibitor for phenoloxidase Phenylthiourea inhibits enzymatic oxidation of DOPA by phenoloxidase (Ki 0 21 M) Phenylthiourea is an effective inhibitor for tyrosinase Phenylthiourea can lead to graying of hair in black rats due to the interference with melanin formation[1][2]