Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioxylenol (mixture of isomers) 90.0+%, TCI America™
SDP

Catalog No. T022625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T022625G Supplier TCI America Supplier No. T022625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25550-52-1
Molecular Formula C8H10S
MDL Number MFCD00010019

Specifications

Color Yellow
Quantity 25 g
Formula Weight 138.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Thioxylenol (mixture of isomers)
EINECSNumber (3)-1094
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.