Learn More
Eurofins Discoverx THIQ, 1 mg. (92-1278)
Supplier:  Eurofins Discoverx 921278
Encompass
THIQ is a synthetic drug molecule and selective agonist for the melanocortin receptor MC4 over MC3 and MC1 (IC50=1.2, 761, and 2067 nM, respectively). THIQ stimulates sexual activity, including enhancement in intracavernosal pressure, but has little effect on appetite or inflammation. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9795