Learn More
TARGETMOL CHEMICALS INC THIQ 10MG
SDP
Catalog No. 502579066
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. THIQ is a selective melanocortin-4 receptor (MC4R) agonist (hMC4R and rMC4R with IC50 of 1.2 nM and 0.6 nM respectively). purity: 98%
Catalog No. 50-257-9066 Supplier TARGETMOL CHEMICALS INC Supplier No. T848510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. THIQ is a selective melanocortin-4 receptor (MC4R) agonist (hMC4R and rMC4R with IC50 of 1.2 nM and 0.6 nM respectively). purity: 98%