Learn More
Teledyne ISCO, Inc Ld Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.00 - 36.00 Ml/Min.; 6,000 Psi
Supplier:  Teledyne ISCO, Inc LD036SRX
Encompass
LD Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 36.00 mL/min.; 6,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9615
Add to Cart
Request a Quote