Learn More
Bachem N-(2-Carbamoyl-ethyl)-Val-Leu-anilide
SDP
Supplier:  Bachem 4025471.0050
Encompass_Preferred
N-(2-Carbamoyl-ethyl)-Val-Leu-anilide, 282725-67-1, C20H32N4O3, Mr 376.50, 50mg. This acrylamide adduct was used as a standard compound in a study on the adduct formation of hemoglobin with acrylamide and N-methylolacrylamide.
Catalog No. 50-194-6940