Learn More
Bachem H-Glu(Phe-OH)-OH
SDP
Supplier:  Bachem 4002375.1000
Encompass_Preferred
H-Glu(Phe-OH)-OH, 7432-24-8, C14H18N2O5, Mr 294.31, 1g. H-g-Glu-Phe-OH. This branched dipeptide was detected in the urine of patients with phenylketonuria and later on in the urine of healthy individuals. g-Glu-Phe and other branched dipeptides have been isolated from hard cheese, where they contribute to the typical flavor of this dairy product.
Catalog No. 50-194-6849