Learn More
Bachem Cyclo(-Leu-Phe)
SDP
Supplier:  Bachem 4025585.1000
Encompass_Preferred
Cyclo(-Leu-Phe), 7280-77-5, C15H20N2O2, Mr 260.34, 1g. This diketopiperazine is converted into the antibacterial dehydropeptide albonoursin by cyclo(Leu-Phe) oxidase from S. albulus KO-23.
Catalog No. 50-194-7456