Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit NexION 5000 Configuration - White/Green
Catalog No. 502016419
Encompass_Preferred
This kit includes: CTFE Barbed Fittings, Small Female Qty.1 (N0777829) PeriPump Tubing Starter Kit (N8150060) Plasma Light Shield(N8152377) PFA Nebulizer (N8152378) Spray Chamber Light Shield (N8152425) Quartz Fixed Torch (N8152428) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) PFA-ST Nebulizer Sample Introduction Line (N8152455) Spray Chamber Drain Line 1/4-28 Fitting (N8152456) 0.25 mm ID Self-aspirating Probe 80 cm (N8152512) 0.30 mm ID Self-aspirating Probe 80 cm (N8152513) SilQ Spray Chamber with Matrix Gas Port (N8152539) White Torch Cassette with Green Stripe (N8161009)
Catalog No. 50-201-6419 Supplier Perkin Elmer US LLC Supplier No. N8160020
Add to Cart
Add to Cart
Request a Quote
This kit includes: CTFE Barbed Fittings, Small Female Qty.1 (N0777829) PeriPump Tubing Starter Kit (N8150060) Plasma Light Shield(N8152377) PFA Nebulizer (N8152378) Spray Chamber Light Shield (N8152425) Quartz Fixed Torch (N8152428) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) PFA-ST Nebulizer Sample Introduction Line (N8152455) Spray Chamber Drain Line 1/4-28 Fitting (N8152456) 0.25 mm ID Self-aspirating Probe 80 cm (N8152512) 0.30 mm ID Self-aspirating Probe 80 cm (N8152513) SilQ Spray Chamber with Matrix Gas Port (N8152539) White Torch Cassette with Green Stripe (N8161009)