Learn More
Perkin Elmer US LLC KBr Window - 4 mm, Qty. 1
Catalog No. 502016436
Encompass_Preferred
This window is made of potassium bromide, is 25 mm in diameter, and has a thickness of 4 mm.
Catalog No. 50-201-6436 Supplier Perkin Elmer US LLC Supplier No. N9302638
This window is made of potassium bromide, is 25 mm in diameter, and has a thickness of 4 mm.