Learn More
Bachem Azido-PEG3-FLAG Trifluoroacetate
SDP
Supplier:  Bachem 4099640.0500
Encompass_Preferred
Azido-PEG3-FLAG Trifluoroacetate, Azido-PEG3-propionyl-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-OH, 2243206-95-1, C50H75N13O24, Mr 1242.22, 0.5mg. This product is used for the attachment of the FLAG-tag by copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC), the prototypical reaction of click chemistry, as described for example by Wang and coworkers. In addition, Azido-PEG3-FLAG contains a PEG-linker. The resulting FLAG-tagged molecules can be detected or purified immunologically, using dye labeled, enzyme coupled or, for purification, immobilized anti-FLAG antibodies.
Catalog No. 50-194-7398