Learn More
Bachem Osteostatin (1-5) amide (human, bovine, dog, horse, mouse, rabbit, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4025761.0025
Encompass_Preferred
Osteostatin (1-5) amide (human, bovine, dog, horse, mouse, rabbit, rat) Trifluoroacetate, H-Thr-Arg-Ser-Ala-Trp-NH2, 155918-12-0, C27H42N10O7, Mr 618.69, 25mg. pTH-rP (107-111) amide (human, bovine, dog, horse, mouse, rabbit, rat), pTH-Related Protein (107-111) amide (human, bovine, dog, horse, mouse, rabbit, rat), Hypercalcemia of Malignancy Factor (107-111) amide (human, bovine, dog, horse, mouse, rabbit, rat). This pTHrP fragment stimulated membrane-associated protein kinase C in freshly isolated rat spleen lymphocytes and thus raises the possibility of being a physiological regulator of the proliferation and other activities of lymphocytes.
Catalog No. 50-194-7084