Learn More
Hach Company Tube, Thistle
Supplier:  Hach Company 2265200
Encompass
Borosilicate glass thistle tube with 20 to 400 screw-thread finish. Used on Hach Distillation Apparatus.
Catalog No. 50-199-5673