Learn More
Cayman Chemical THJ2201 N-(5-hydroxypentyl) metabolite, Purity: ≥98%, CAS: 1850409-16-3, 5 mg
SDP
Catalog No. 502895663
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of THJ2201 and THJ 018; intended for research and forensic applications
Catalog No. 50-289-5663 Supplier Cayman Chemical Supplier No. 90037365 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of THJ2201 and THJ 018; intended for research and forensic applications