Learn More
Medchemexpress LLC Thk-5117 | 1374107-54-6 | 98.5% | 326.36 | C19H19FN2O2 | 5 MG
SDP
Catalog No. 5000212127
Encompass_Preferred
THK-5117 is an arylquinoline research compound used as a tau protein-binding agent for laboratory studies and imaging probe development. It is supplied as a high-purity solid and is intended for research use, with recommended storage under inert atmosphere and at low temperatures to preserve stability.
  • High purity suitable for research applications.
  • Molecular weight 326.36 for accurate mass calculations.
  • Available in small, lab-friendly quantities for assay development.
  • Stable when stored under nitrogen at recommended temperatures.
  • Includes data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-12127 Supplier Medchemexpress LLC Supplier No. HY1011825MG
THK-5117 is an arylquinoline research compound used as a tau protein-binding agent for laboratory studies and imaging probe development. It is supplied as a high-purity solid and is intended for research use, with recommended storage under inert atmosphere and at low temperatures to preserve stability.
  • High purity suitable for research applications.
  • Molecular weight 326.36 for accurate mass calculations.
  • Available in small, lab-friendly quantities for assay development.
  • Stable when stored under nitrogen at recommended temperatures.
  • Includes data sheet, certificate of analysis, and safety data sheet.