Learn More
Medchemexpress LLC Thk-5117 | 1374107-54-6 | 98.5% | 326.36 g/mol | C19H19FN2O2 | 50 MG
SDP
Catalog No. 5000213370
Encompass_Preferred
THK-5117 is an arylquinoline research compound that binds tau fibrils with high affinity (Ki ≈ 10.5 nM). It is used in preclinical studies of tau pathology and in PET radioligand development. The compound has CAS 1374107-54-6, molecular weight 326.36 g/mol, and is supplied as a 50 mg research vial.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • Suitable for preclinical tau imaging and tracer development.
  • High chemical purity (≈98.5%).
  • Supplied as a 50 mg research-grade vial.
  • Store under inert gas at 4°C; in solvent keep at -80°C for long-term storage.
Catalog No. 50-002-13370 Supplier Medchemexpress LLC Supplier No. HY10118250MG
THK-5117 is an arylquinoline research compound that binds tau fibrils with high affinity (Ki ≈ 10.5 nM). It is used in preclinical studies of tau pathology and in PET radioligand development. The compound has CAS 1374107-54-6, molecular weight 326.36 g/mol, and is supplied as a 50 mg research vial.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • Suitable for preclinical tau imaging and tracer development.
  • High chemical purity (≈98.5%).
  • Supplied as a 50 mg research-grade vial.
  • Store under inert gas at 4°C; in solvent keep at -80°C for long-term storage.