Learn More
Electron Microscopy Sciences Athene Thick and Thin Bar Grid with Center Mark, 200 mesh Cu
SDP
Catalog No. 501926676
Encompass_Preferred
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Copper
Catalog No. 50-192-6676 Supplier Electron Microscopy Sciences Supplier No. ATT200CU
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Copper