Learn More
Hach Company THM Reagent 3 Solution, 100 mL
Supplier:  Hach Company 2754142
Encompass
For determination of Total Trihalomethanes by the THM Plus™ Method. Hach Method 10132. Part of Reagent Set 2790800. 100 mL Bottle
Catalog No. 50-199-6626