Learn More
Andwin Scientific THML SEAL FILM N/S 100/PK
SDP
Catalog No. NC9784612
Encompass_Preferred
The ThermalSeal 2 Sealing Film is ideal for classic thermal cycling applications. This product consists of a 50 m polyester film with a 41 m layer of acrylic adhesive. Compared to previous models, this film provide better adhesion, less evaporation, longer end tabs for easier handling, and less tendency to tear during removal from the plate. This film measures 79.4 x 139.7 mm, which is enough to securely seal all the area of PCR plates. It is sold in a pack of 100.
Catalog No. NC9784612 Supplier Andwin Scientific Supplier No. 100THERPLT
The ThermalSeal 2 Sealing Film is ideal for classic thermal cycling applications. This product consists of a 50 m polyester film with a 41 m layer of acrylic adhesive. Compared to previous models, this film provide better adhesion, less evaporation, longer end tabs for easier handling, and less tendency to tear during removal from the plate. This film measures 79.4 x 139.7 mm, which is enough to securely seal all the area of PCR plates. It is sold in a pack of 100.