Learn More
Electron Microscopy Sciences Athene Thin Bar Grid with Center Mark, 300 mesh Grid, Ni
SDP
Catalog No. 501926674
Encompass_Preferred
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Nickel
Catalog No. 50-192-6674 Supplier Electron Microscopy Sciences Supplier No. AT300NI
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Nickel