Learn More
Norell Inc THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, (3-12mm INSIDE DIAMETER), 12 in. LENGTH, PACKED IN LOT OF 9 TUBES
SDP
Catalog No. 50821129
Encompass_Preferred
THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, (3-12mm INSIDE DIAMETER), 12 in. LENGTH, PACKED IN LOT OF 9 TUBES
Catalog No. 50-821-129 Supplier Norell Inc Supplier No. TWT312
THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, (3-12mm INSIDE DIAMETER), 12 in. LENGTH, PACKED IN LOT OF 9 TUBES