Learn More
Norell Inc THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, 7.68mm OD X 7mm ID, 12 in. LENGTH, PACKED IN LOTS OF 4
SDP
Catalog No. 50821124
Encompass_Preferred
THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, 7.68mm OD X 7mm ID, 12 in. LENGTH, PACKED IN LOTS OF 4
Catalog No. 50-821-124 Supplier Norell Inc Supplier No. TWT7
THIN-WALL TRANSPARENT FLUOROPOLYMER FEP TUBING, 7.68mm OD X 7mm ID, 12 in. LENGTH, PACKED IN LOTS OF 4