Learn More
Aviva Systems Biology Corp THOC1 Antibody - C-terminal region (ARP40578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THOC1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human THOC1. Alternate names: HPR1, P84, P84N5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53209