Learn More
Aviva Systems Biology Corp THOC4 Antibody - N-terminal region (ARP43055_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43055_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against THOC4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human THOC4. Alternate names: ALY, BEF, REF, THOC4, ALY/REFApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58410