Learn More
Thomas Scientific Thomas Scientific Deep Well Plate, 96-Well, Angular Cavities, Polypropylene, 2.5 mL
SDP
Catalog No. NC3981200
Encompass
This deep well plate features 96 wells with angular cavities, constructed from highly transparent polypropylene, offering a 2.5 mL capacity per well. Its design ensures compatibility with standard laboratory equipment and provides reliable sample handling and identification.
  • Made of highly transparent polypropylene, ensuring an optimal surface for microscopic analysis.
  • Standardized dimensions (128 × 85 mm) for compatibility with most common laboratory machines.
  • Available in different shapes: U- and V-shape.
  • Alphanumeric coding for clear sample assignment and identification.
Catalog No. NC3981200 Supplier Thomas Scientific Supplier No. 23A00D096
This deep well plate features 96 wells with angular cavities, constructed from highly transparent polypropylene, offering a 2.5 mL capacity per well. Its design ensures compatibility with standard laboratory equipment and provides reliable sample handling and identification.
  • Made of highly transparent polypropylene, ensuring an optimal surface for microscopic analysis.
  • Standardized dimensions (128 × 85 mm) for compatibility with most common laboratory machines.
  • Available in different shapes: U- and V-shape.
  • Alphanumeric coding for clear sample assignment and identification.