Learn More
Electron Microscopy Sciences Thompson Monolayer Embedding Mold
SDP
Catalog No. 50930544
Encompass_Preferred
This mold used for embedding mammalian cells in monolayer culture in preparation of sectioning for light or electron microscopy. Tissue culture monolayer embedding molds eliminate the need for traditional BEEM capsules. The improved molds can be used to transfer the majority of cells from the entire surface of a cover slip to avoid losses of large amounts of cellular information occurring only randomly. In addition, other problems such as surface blemishes caused by air entrapment associated with BEEM capsules are eliminated.
Catalog No. 50-930-544 Supplier Electron Microscopy Sciences Supplier No. 70924
This mold used for embedding mammalian cells in monolayer culture in preparation of sectioning for light or electron microscopy. Tissue culture monolayer embedding molds eliminate the need for traditional BEEM capsules. The improved molds can be used to transfer the majority of cells from the entire surface of a cover slip to avoid losses of large amounts of cellular information occurring only randomly. In addition, other problems such as surface blemishes caused by air entrapment associated with BEEM capsules are eliminated.