Learn More
Biotium Thomsen-Friedenreich Antigen / CD176 (Pan Carcinoma Marker)(A84-A/F10), 0.2 mg/mL
Supplier:  Biotium BNUB1294500
Encompass_Preferred
Recognizes a disaccharide epitope, Gal 1-3GalNAc, of Thomsen-Friedenreich (TF) antigen. It is specific for both anomeric forms of the disaccharide (TF and TF , including related structures on the glycolipid) and shows no cross-reactivity with sialylated glycophorin. The TF antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. Considered to be a pan-carcinoma marker. It is capable to agglutinate desialylated red blood cells. During metastasis, the ability of malignant cells to form multicellular aggregates via homotypic or heterotypic aggregation and their adhesion to the endothelium are critical. It is involved in tumor cell adhesion and tissue invasion. It also causes an immune response, and overexpression of the antigen causes cancer cells to be more sensitive to natural killer cell lysis. TF antigen is suppressed in normal healthy cells and represents one of the few chemi
Catalog No. 50-006-58668