Learn More
Med Vet International Thomsen’s Universal Dental Float, 20", J0058S
Catalog No. 5000584293
Encompass_Preferred
Float head is hinge attached, making this float universal for upper and lower molars. Float blade is concave from side to side, stays on dental surface. Long lasting tungsten carbide blade.
Catalog No. 50-005-84293 Supplier Med Vet International Supplier No. JOR58S
Float head is hinge attached, making this float universal for upper and lower molars. Float blade is concave from side to side, stays on dental surface. Long lasting tungsten carbide blade.