Learn More
Thomson Instrument Company Thomson Rapid Clear 2mL, 96-Well 0.2µm Filter Plate, Sterile
SDP
Supplier:  Thomson Instrument Company 921746
Encompass_Preferred
Thomson Filter Plates (24- and 96-well) enable fast, parallel plate-to-plate processing for biological and analytical workflows.
• Rapid Clear 0.2 µm depth filtration — multi-layer, graded matrix captures progressively smaller cells, debris, and aggregates for reliable clarification by swing-bucket centrifuge.
• Excels with mammalian cultures — clarifies low- or high-density CHO (stable/transient), HEK293, hybridoma, and other mammalian cell lines.
• High-throughput in ~5 minutes — processes 24 wells in parallel with uniform flow for consistent results; long-neck drip geometry helps limit cross-well carryover.
• Protein-friendly matrix — engineered to minimize interaction with antibodies for strong recovery.
• Automation-ready formats — ANSI/SLAS plates with matching lids/seals.
• Sterile (SAL 10⁻⁶)
• Made in the USA
Catalog No. 50-222-1352