Learn More
Medchemexpress LLC HY-N4084 5mg Medchemexpress, Thonningianin A CAS:271579-11-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40845MG
Encompass_Preferred
Medchemexpress, HY-N4084 5mg Thonningianin A CAS:271579-11-4 Thonningianin A, an ellagitannin, is isolated from the methanolic extract of the African medicinal herb, Thonningia sanguinea. The antioxidant properties of Th A involve radical scavenging, anti-superoxide formation and metal chelation. Anti-cancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2921