Learn More
Apexbio Technology LLC Thonzonium Bromide,50mg CAS# 553-08-2
SDP
Catalog No. 502240158
Encompass_Preferred
Thonzonium Bromide is a surface active agent and an inhibitor of V-ATPase proton transport with EC50 value of 69 µM [1] [2].Vacuolar ATPases (V-ATPases) are proton pumps that maintain pH homeostasis. V-ATPases couple the energy of ATP hydrolysis to proton transport across intracellular membranes [2].Thonzonium Bromide is a surface active agent and an inhibitor of V-ATPase proton transport. In wild-type yeast cells, thonzonium bromide significantly decreased the cytosolic pH to 6.22. In vacuolar membrane vesicles, thonzonium bromide inhibited proton transport with EC50 value of 69 µM in a dose-dependent way. However, thonzonium bromide didn’t inhibit ATP hydrolysis, which suggested that thonzonium bromide uncoupled V-ATPase proton pumps. In wild-type yeast cells, thonzonium bromide (1 µM) didn’t inhibit cell growth. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0158 Supplier Apexbio Technology LLC Supplier No. B496250
Thonzonium Bromide is a surface active agent and an inhibitor of V-ATPase proton transport with EC50 value of 69 µM [1] [2].Vacuolar ATPases (V-ATPases) are proton pumps that maintain pH homeostasis. V-ATPases couple the energy of ATP hydrolysis to proton transport across intracellular membranes [2].Thonzonium Bromide is a surface active agent and an inhibitor of V-ATPase proton transport. In wild-type yeast cells, thonzonium bromide significantly decreased the cytosolic pH to 6.22. In vacuolar membrane vesicles, thonzonium bromide inhibited proton transport with EC50 value of 69 µM in a dose-dependent way. However, thonzonium bromide didn’t inhibit ATP hydrolysis, which suggested that thonzonium bromide uncoupled V-ATPase proton pumps. In wild-type yeast cells, thonzonium bromide (1 µM) didn’t inhibit cell growth. Other sizes are also available. Please inqury us for quote.