Learn More
Medchemexpress LLC Thonzonium bromide | 553-08-2 | 99.7% | 50 MG
SDP
Catalog No. 5000412877
Encompass_Preferred
Thonzonium bromide is an antibacterial and monocationic surface-active agent. It inhibits proton transport and has been shown to inhibit RANKL-induced osteoclast formation and bone resorption in vitro. This compound also prevents LPS-induced bone loss in vivo.
  • Inhibits proton transport
  • Prevents LPS-induced bone loss
  • Blocks activation of NF-κB, ERK, and c-Fos
  • Disrupts F-actin ring formation in mature osteoclasts
  • Exhibits protective effects in murine models
Catalog No. 50-004-12877 Supplier Medchemexpress LLC Supplier No. HYB124650MG
Thonzonium bromide is an antibacterial and monocationic surface-active agent. It inhibits proton transport and has been shown to inhibit RANKL-induced osteoclast formation and bone resorption in vitro. This compound also prevents LPS-induced bone loss in vivo.
  • Inhibits proton transport
  • Prevents LPS-induced bone loss
  • Blocks activation of NF-κB, ERK, and c-Fos
  • Disrupts F-actin ring formation in mature osteoclasts
  • Exhibits protective effects in murine models