Learn More
Medchemexpress LLC Thonzonium bromide | 553-08-2 | 99.86% | 1 ML
SDP
Catalog No. 5000412870
Encompass_Preferred
Thonzonium bromide is an antibacterial agent and monocationic surface-active agent that inhibits RANKL-induced osteoclast formation and bone resorption in vitro. It prevents LPS-induced bone loss in vivo by blocking RANKL-induced activation of NF-κB, ERK, c-Fos, and NFATc1, which are crucial for osteoclast formation. Additionally, it disrupts F-actin ring formation in mature osteoclasts and inhibits proton transport in a dose-dependent manner.
  • Inhibits RANKL-induced osteoclast formation
  • Prevents LPS-induced bone loss in vivo
  • Blocks key signaling pathways for osteoclast formation
  • Disrupts F-actin ring formation in mature osteoclasts
  • Inhibits proton transport
Catalog No. 50-004-12870 Supplier Medchemexpress LLC Supplier No. HYB124610MM/1ML
Thonzonium bromide is an antibacterial agent and monocationic surface-active agent that inhibits RANKL-induced osteoclast formation and bone resorption in vitro. It prevents LPS-induced bone loss in vivo by blocking RANKL-induced activation of NF-κB, ERK, c-Fos, and NFATc1, which are crucial for osteoclast formation. Additionally, it disrupts F-actin ring formation in mature osteoclasts and inhibits proton transport in a dose-dependent manner.
  • Inhibits RANKL-induced osteoclast formation
  • Prevents LPS-induced bone loss in vivo
  • Blocks key signaling pathways for osteoclast formation
  • Disrupts F-actin ring formation in mature osteoclasts
  • Inhibits proton transport