Learn More
Medchemexpress LLC Thonzylamine | 91-85-0 | 98.1% | 25 MG
SDP
Catalog No. 5000414010
Encompass_Preferred
Thonzylamine is an orally active H1 histamine receptor antagonist with good antihistaminic and antianaphylactic properties. It is used for the research of various hypersensitivity diseases, nasal congestion, and allergic conjunctivitis.
  • Orally active H1 histamine receptor antagonist
  • Good antihistaminic and antianaphylactic properties
  • Suitable for research on hypersensitivity diseases
  • Applicable for studying nasal congestion
  • Used in research for allergic conjunctivitis and other allergic diseases
Catalog No. 50-004-14010 Supplier Medchemexpress LLC Supplier No. 25MGHYB1317
Thonzylamine is an orally active H1 histamine receptor antagonist with good antihistaminic and antianaphylactic properties. It is used for the research of various hypersensitivity diseases, nasal congestion, and allergic conjunctivitis.
  • Orally active H1 histamine receptor antagonist
  • Good antihistaminic and antianaphylactic properties
  • Suitable for research on hypersensitivity diseases
  • Applicable for studying nasal congestion
  • Used in research for allergic conjunctivitis and other allergic diseases