Learn More
THORLABS INC Thorlabs Aspheric Lens, 25.4 mm Diameter, 18.39 mm EFL, 0.85 NA, S-LAH64
Catalog No. NC3985410
Encompass
This aspheric lens is designed for use in the 650 to 1050 nm range. It is fabricated from S-LAH64 glass and has a numerical aperture of 0.85. The aspheric design of this lens eliminates spherical aberration, making it ideal for a variety of applications, including collimating the output of a laser diode, coupling light into an optical fiber, and focusing a collimated beam.
  • Precision molded glass aspheric lenses
  • Diffraction limited performance for collimation and focusing applications
  • Available unmounted or mounted in an SM1-threaded (1.035″-40) mount
  • Coating options for 350 to 500 nm, 600 to 1050 nm, 1050 to 1700 nm, or 400 to 1200 nm
  • Eliminates spherical aberration
  • Designed for optimal performance with C-mount and T-mount laser diode packages
  • Custom aspheric lenses available
  • Compatible with ASR series aspheric lens mounts and AD15F adapter
Catalog No. NC3985410 Supplier THORLABS INC Supplier No. LJ4147RMA
This aspheric lens is designed for use in the 650 to 1050 nm range. It is fabricated from S-LAH64 glass and has a numerical aperture of 0.85. The aspheric design of this lens eliminates spherical aberration, making it ideal for a variety of applications, including collimating the output of a laser diode, coupling light into an optical fiber, and focusing a collimated beam.
  • Precision molded glass aspheric lenses
  • Diffraction limited performance for collimation and focusing applications
  • Available unmounted or mounted in an SM1-threaded (1.035″-40) mount
  • Coating options for 350 to 500 nm, 600 to 1050 nm, 1050 to 1700 nm, or 400 to 1200 nm
  • Eliminates spherical aberration
  • Designed for optimal performance with C-mount and T-mount laser diode packages
  • Custom aspheric lenses available
  • Compatible with ASR series aspheric lens mounts and AD15F adapter