Learn More
THORLABS INC Thorlabs Axicon, 1.0°, Uncoated, UV Fused Silica, 1 in.
Catalog No. NC3985438
Encompass
This axicon is designed with a 1.0° angle and is constructed from UV Fused Silica. It features an uncoated surface and has a 1 in. diameter.
  • Features a 1.0° angle
  • Uncoated surface for broad spectral transmission
  • Constructed from UV Fused Silica for ultraviolet applications
  • Measures 1 in. (25.4 mm) in diameter
Catalog No. NC3985438 Supplier THORLABS INC Supplier No. AX251
This axicon is designed with a 1.0° angle and is constructed from UV Fused Silica. It features an uncoated surface and has a 1 in. diameter.
  • Features a 1.0° angle
  • Uncoated surface for broad spectral transmission
  • Constructed from UV Fused Silica for ultraviolet applications
  • Measures 1 in. (25.4 mm) in diameter