Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THORLABS INC Thorlabs Broad Spectrum LED, 300 to 1000 nm, 5 mW Output, M3 Taps

Catalog No. NC3985226
Encompass

This compact, stable, low-cost Broad Spectrum LED provides 300 to 1000 nm wavelength range (low output below 350 nm) for spectroscopy, generated by a phosphor-coated blue LED. It has a Ø1.5″ (38.1 mm) body with integrated heatsink and tapped holes for mounting, ideal for compact spectrometers or standalone use.

  • Broad spectrum light emitting diode
  • Wavelength range: 300 to 1000 nm
  • Minimum output power: 5 mW
  • Low thermal drift with integrated heatsink
  • Ø1.5″ (38.1 mm) body with 4-40 and M3 tapped holes
  • Lifetime (MTTF): >10,000 hrs
  • Operating voltage: 3.0 to 4.5 VDC
  • Operating current: 600 mA max.
  • Operating temperature: 0°C to 40°C
  • Housing dimensions: Ø1.5″ x 2.22″ (38.1 mm x 56.4 mm)
  • Weight: 0.18 lbs (0.08 kg)
  • Not RoHS compliant

Catalog No. NC3985226 Supplier THORLABS INC Supplier No. LSB03B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compact, stable, low-cost Broad Spectrum LED provides 300 to 1000 nm wavelength range (low output below 350 nm) for spectroscopy, generated by a phosphor-coated blue LED. It has a Ø1.5″ (38.1 mm) body with integrated heatsink and tapped holes for mounting, ideal for compact spectrometers or standalone use.

  • Broad spectrum light emitting diode
  • Wavelength range: 300 to 1000 nm
  • Minimum output power: 5 mW
  • Low thermal drift with integrated heatsink
  • Ø1.5″ (38.1 mm) body with 4-40 and M3 tapped holes
  • Lifetime (MTTF): >10,000 hrs
  • Operating voltage: 3.0 to 4.5 VDC
  • Operating current: 600 mA max.
  • Operating temperature: 0°C to 40°C
  • Housing dimensions: Ø1.5″ x 2.22″ (38.1 mm x 56.4 mm)
  • Weight: 0.18 lbs (0.08 kg)
  • Not RoHS compliant

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.