Learn More
THORLABS INC Thorlabs Broadband Dielectric Mirror, 1 in., 750 to 1100 nm, Fused Silica, 10-pack
Supplier: THORLABS INC BB1E0310
Thorlabs' Fused Silica Broadband Dielectric Mirrors are designed to offer excellent reflectance. This particular offering is a 10-pack of 1 in. diameter mirrors with a 750 to 1100 nm wavelength range.
- Broadband dielectric mirror
- 750 to 1100 nm wavelength range
- 10-pack configuration
- Fused silica substrate material
- Reflectivity: Ravg > 99% for S- and P-polarization for angles of incidence from 0° to 45°
- Designed for CW and nanosecond lasers