Learn More
THORLABS INC Thorlabs Broadband Dielectric Mirror, 1 in. Diameter, 400 to 750 nm Range, 10 Pack
Catalog No. NC3985254
Encompass
This Thorlabs Broadband Dielectric Mirror features a 1 in. diameter and a wavelength range of 400 to 750 nm. It is available in a convenient 10 pack and is both RoHS and REACH compliant, offering a reliable solution for various optical applications.
  • 1 in. diameter broadband dielectric mirror
  • Wavelength range of 400 to 750 nm
  • Available in a convenient 10 pack
  • RoHS compliant
  • REACH compliant
  • Backed by a two-year warranty
Catalog No. NC3985254 Supplier THORLABS INC Supplier No. BB1E0210
This Thorlabs Broadband Dielectric Mirror features a 1 in. diameter and a wavelength range of 400 to 750 nm. It is available in a convenient 10 pack and is both RoHS and REACH compliant, offering a reliable solution for various optical applications.
  • 1 in. diameter broadband dielectric mirror
  • Wavelength range of 400 to 750 nm
  • Available in a convenient 10 pack
  • RoHS compliant
  • REACH compliant
  • Backed by a two-year warranty