Learn More
THORLABS INC Thorlabs Cage Rotation Mount, Ø1″ Optics, SM1 Threaded, 8-32 Tap
Supplier: THORLABS INC CRM1T
This is a cage rotation mount designed for Ø1″ optics. It features SM1 threading and an 8-32 tap, making it suitable for integrating Ø1″ or Ø25.0 mm optics into a 30 mm cage system.
- Designed for Ø1″ or Ø25.0 mm optics.
- Features SM1 threading.
- Equipped with an 8-32 tap.
- Suitable for 30 mm cage systems.