Learn More
THORLABS INC Thorlabs Clamping Forks and Post Holders Kit, Imperial and Universal Components, 130 Pieces
Catalog No. NC3985227
Encompass
This clamping forks and post holders kit is a comprehensive collection of 130 essential optical mounting components. It is designed to provide convenient, organized storage and easy access to a variety of clamping forks, as well as universal and pedestal post holders. The heavy-duty, welded steel frame ensures durability, and its versatile design allows for wall mounting or stable stacking.
  • Contains 130 popular optical mounting components.
  • Includes clamping forks and universal and pedestal post holders.
  • Features a heavy-duty, welded steel cabinet frame.
  • Cabinet can be wall mounted or stably stacked.
  • Each compartment is labeled with an image and item number for easy identification.
  • Equipped with extra dividers and white stickers for custom labeling.
  • RoHS Compliant and REACH Compliant.
Catalog No. NC3985227 Supplier THORLABS INC Supplier No. ESK14
This clamping forks and post holders kit is a comprehensive collection of 130 essential optical mounting components. It is designed to provide convenient, organized storage and easy access to a variety of clamping forks, as well as universal and pedestal post holders. The heavy-duty, welded steel frame ensures durability, and its versatile design allows for wall mounting or stable stacking.
  • Contains 130 popular optical mounting components.
  • Includes clamping forks and universal and pedestal post holders.
  • Features a heavy-duty, welded steel cabinet frame.
  • Cabinet can be wall mounted or stably stacked.
  • Each compartment is labeled with an image and item number for easy identification.
  • Equipped with extra dividers and white stickers for custom labeling.
  • RoHS Compliant and REACH Compliant.