Learn More
THORLABS INC Thorlabs Cotton Optic Gloves, Large, 12 Pairs Per Package
Catalog No. NC3985292
Encompass
These disposable cotton gloves are designed for handling optics and can be worn underneath latex gloves as a glove liner. They are packaged in bags containing 12 pairs of cotton gloves.
  • Glove size: Large
  • Palm size circumference around the palm not including the thumb: 8.75 in. to 9.5 in. (22.23 cm to 24.13 cm)
  • Maximum middle finger length: 3.75 in. (9.53 cm)
  • 12 pairs per package
  • Suitable for handling optics
  • Can be worn underneath latex gloves as a glove liner
Catalog No. NC3985292 Supplier THORLABS INC Supplier No. MC6L
These disposable cotton gloves are designed for handling optics and can be worn underneath latex gloves as a glove liner. They are packaged in bags containing 12 pairs of cotton gloves.
  • Glove size: Large
  • Palm size circumference around the palm not including the thumb: 8.75 in. to 9.5 in. (22.23 cm to 24.13 cm)
  • Maximum middle finger length: 3.75 in. (9.53 cm)
  • 12 pairs per package
  • Suitable for handling optics
  • Can be worn underneath latex gloves as a glove liner