Learn More
THORLABS INC THORLABS Ground Glass Diffuser, 1 in., Protected Silver Reflective, 120 Grit
Catalog No. NC3985377
Encompass
This ground glass diffuser features a 1 in. diameter and a 120 grit finish, enhanced with a protected silver reflective coating. Designed for various optical applications, this product offers reliable light diffusion performance.
  • Protected silver reflective coating for enhanced durability and performance.
  • Ground glass construction provides excellent light diffusion.
  • 120 grit finish ensures consistent diffusion characteristics.
  • 1 in. diameter for standard optical setups.
  • RoHS compliant for environmental safety.
Catalog No. NC3985377 Supplier THORLABS INC Supplier No. DG10120P01
This ground glass diffuser features a 1 in. diameter and a 120 grit finish, enhanced with a protected silver reflective coating. Designed for various optical applications, this product offers reliable light diffusion performance.
  • Protected silver reflective coating for enhanced durability and performance.
  • Ground glass construction provides excellent light diffusion.
  • 120 grit finish ensures consistent diffusion characteristics.
  • 1 in. diameter for standard optical setups.
  • RoHS compliant for environmental safety.