Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THORLABS INC THORLABS HeNe Laser, 632.8 nm, 10 mW, Fluctuating Polarization

Catalog No. NC3985414
Encompass

This HeNe laser provides a stable 632.8 nm output with 10 mW power and fluctuating polarization. It includes a 100 to 240 VAC power supply, making it suitable for various laboratory and scientific applications.

  • Wavelength: 632.8 nm
  • Output power: 10 mW
  • Polarization: Fluctuating
  • Power supply: 100 to 240 VAC power supply included
  • RoHS exempt — Annex III — 7(c)-I
  • REACH SVHC > 0.1% lead
  • One year warranty

Catalog No. NC3985414 Supplier THORLABS INC Supplier No. HNL100RB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This HeNe laser provides a stable 632.8 nm output with 10 mW power and fluctuating polarization. It includes a 100 to 240 VAC power supply, making it suitable for various laboratory and scientific applications.

  • Wavelength: 632.8 nm
  • Output power: 10 mW
  • Polarization: Fluctuating
  • Power supply: 100 to 240 VAC power supply included
  • RoHS exempt — Annex III — 7(c)-I
  • REACH SVHC > 0.1% lead
  • One year warranty

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.