Learn More
THORLABS INC THORLABS INC Cleaning Kit, Red First Contact Polymer, for Optics, RoHS Compliant
Catalog No. NC3985310
Encompass
This cleaning kit utilizes Red First Contact™ Polymer technology and is specifically designed for cleaning optics. The kit has a package weight of 0.83 kg.
  • Features First Contact™ Polymer technology
  • Designed for optics cleaning
  • Comes with a two-year warranty; incorporated light sources are warrantied for the lesser of one year or the number of hours stated in specifications
  • RoHS compliant
  • REACH compliant
Catalog No. NC3985310 Supplier THORLABS INC Supplier No. PCK3
This cleaning kit utilizes Red First Contact™ Polymer technology and is specifically designed for cleaning optics. The kit has a package weight of 0.83 kg.
  • Features First Contact™ Polymer technology
  • Designed for optics cleaning
  • Comes with a two-year warranty; incorporated light sources are warrantied for the lesser of one year or the number of hours stated in specifications
  • RoHS compliant
  • REACH compliant