Learn More
THORLABS INC THORLABS INC Mirror Mount, Ø1″ Optic, Left-Handed, Stainless Steel
Catalog No. NC3985275
Encompass
This left-handed Polaris™ mirror mount provides extreme stability for Ø1″ (Ø25 mm) optics. It features a monolithic stainless steel body, an all-stainless-steel adjustment mechanism, and flexure pivots that allow for smooth, continuous motion. This design inhibits the “creeping” and “walking” that can occur with conventional multi-piece mirror mounts and provides a pure tilt. All Polaris mounts are designed for a 10 arcsec (or better) pointing stability over a 12-hour period.
  • Monolithic stainless steel body
  • All stainless steel adjustment screws
  • Thermally matched actuators
  • Ø1″ (Ø25 mm) optic compatibility
  • Left-handed design
  • Pure kinematic motion
  • Flexure actuated
  • Two M4 tapped holes
Catalog No. NC3985275 Supplier THORLABS INC Supplier No. MC10BM
This left-handed Polaris™ mirror mount provides extreme stability for Ø1″ (Ø25 mm) optics. It features a monolithic stainless steel body, an all-stainless-steel adjustment mechanism, and flexure pivots that allow for smooth, continuous motion. This design inhibits the “creeping” and “walking” that can occur with conventional multi-piece mirror mounts and provides a pure tilt. All Polaris mounts are designed for a 10 arcsec (or better) pointing stability over a 12-hour period.
  • Monolithic stainless steel body
  • All stainless steel adjustment screws
  • Thermally matched actuators
  • Ø1″ (Ø25 mm) optic compatibility
  • Left-handed design
  • Pure kinematic motion
  • Flexure actuated
  • Two M4 tapped holes